2-Iodo-6-nitro-4-[(trifluoromethyl)thio]phenol 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Iodo-6-nitro-4-[(trifluoromethyl)sulphanyl]phenol, 4-Hydroxy-3-iodo-5-nitrophenyl trifluoromethyl sulphide

PC49360-1G PC49360-250MG
APOSPC49360-1GEA 5670 DKK
APOSPC49360-1G APOSPC49360-250MG
2-Iodo-6-nitro-4-[(trifluoromethyl)thio]phenol 95%
2-Iodo-6-nitro-4-[(trifluoromethyl)thio]phenol
Formel: C₇H₃F₃INO₃S
Opbevaringstemperatur: Køleskab
MDL Number: MFCD24850032
CAS-nummer: 1440535-09-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR