3-Fluor-5-nitrobenzoesyre 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC48725-5G
APOSPC48725-5GEA 719 DKK
APOSPC48725-5G
3-Fluor-5-nitrobenzoesyre 97%
3-Fluor-5-nitrobenzoesyre
Formel: C₇H₄FNO₄
Opbevaringstemperatur: Fryser
MDL Number: MFCD05864360
CAS-nummer: 14027-75-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR