Ethyl 2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butyrate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Ethyl 4,4,4,4',4',4'-hexafluoro-DL-valinate, 4,4,4,4',4',4'-Hexafluorovaline, ethyl ester

PC450233-1G PC450233-5G
APOSPC450233-1GEA 589 DKK
APOSPC450233-1G APOSPC450233-5G
Ethyl 2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butyrate
Ethyl-2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butyrate
Formel: C₇H₉F₆NO₂
Kogepunkt: 70…72 °C/10mmHg
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00040845
CAS-nummer: 78185-92-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR