5-(Methoxycarbonyl)-1-(4-fluorobenzyl)-1H-pyrrolo[2,3-c]pyridine-3-carboxylic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC430584-100MG
APOSPC430584-100MGEA 4260 DKK
APOSPC430584-100MG
5-(Methoxycarbonyl)-1-(4-fluorobenzyl)-1H-pyrrolo[2,3-c]pyridine-3-carboxylic acid
5-(Methoxycarbonyl)-1-(4-fluorobenzyl)-1H-pyrrolo[2,3-c]pyridine-3-carboxylic acid
Formel: C₁₇H₁₃FN₂O₄
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD14585350
CAS-nummer: 932396-34-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR