3-[4-(Trifluoromethyl)phenyl]propanal

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-[4-(Trifluoromethyl)phenyl]propionaldehyde, 4-(3-Oxoprop-1-yl)benzotrifluoride

PC4059-100MG PC4059-250MG
APOSPC4059-100MGEA 480 DKK
APOSPC4059-100MG APOSPC4059-250MG
3-[4-(Trifluoromethyl)phenyl]propanal
3-[4-(Trifluoromethyl)phenyl]propanal
Formel: C₁₀H₉F₃O
Flammepunkt: 96 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07772919
CAS-nummer: 166947-09-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR