2-Nitro-5-(trifluoromethyl)aniline

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-Amino-4-nitrobenzotrifluoride

PC3972-1G PC3972-5G PC3972-10G
APOSPC3972-1GEA 654 DKK
APOSPC3972-1G APOSPC3972-5G APOSPC3972-10G
2-Nitro-5-(trifluoromethyl)aniline
2-Nitro-5-(trifluoromethyl)aniline
Formel: C₇H₅F₃N₂O₂
Smeltepunkt: 98…102 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00042447
CAS-nummer: 402-14-2
UN nummer: 2811
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR