3-Fluoro-2-methylbenzotrifluoride 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

PC3319-1G PC3319-25G PC3319-5G
APOSPC3319-1GEA 164 DKK
APOSPC3319-1G APOSPC3319-25G APOSPC3319-5G
3-Fluoro-2-methylbenzotrifluoride 98%
3-Fluoro-2-methylbenzotrifluoride
Formel: C₈H₆F₄
Massefylde: 1,2 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08062608
CAS-nummer: 910911-43-2
UN nummer: 1993
ADR: 3,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR