4-(Difluoromethyl)-2-(trifluoromethyl)benzonitrile 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

PC32953-1G PC32953-250MG PC32953-5G
APOSPC32953-1GEA 567 DKK
APOSPC32953-1G APOSPC32953-250MG APOSPC32953-5G
4-(Difluoromethyl)-2-(trifluoromethyl)benzonitrile 95%
4-(Difluoromethyl)-2-(trifluoromethyl)benzonitrile
Formel: C₉H₄F₅N
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD28790571
CAS-nummer: 1807192-90-0
UN nummer: 3265
ADR: 8,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR