2,4-Difluor-3-methoxyanilin 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-Amino-2,6-difluoroanisole

PC303017-10G PC303017-5G PC303017-1G
APOSPC303017-10GEA 3550 DKK
APOSPC303017-10G APOSPC303017-5G APOSPC303017-1G
2,4-Difluor-3-methoxyanilin 98%
2,4-Difluor-3-methoxyanilin
Formel: C₇H₇F₂NO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04115929
CAS-nummer: 886499-08-7
UN nummer: 2810
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR