1-[2-(Trifluoromethyl)phenyl]propylamine 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-(1-Aminoprop-1-yl)benzotrifluoride, α-Ethyl-2-(trifluoromethyl)benzylamine

PC2542-1G PC2542-250MG
APOSPC2542-1GEA 4910 DKK
APOSPC2542-1G APOSPC2542-250MG
1-[2-(Trifluoromethyl)phenyl]propylamine 98%
1-[2-(Trifluoromethyl)phenyl]propylamine
Formel: C₁₀H₁₂F₃N
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07784288
CAS-nummer: 473732-54-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR