2-Chlor-4'-fluoracetanilid 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Chloro-N-(4-fluorophenyl)acetamide, N-(Chloroacetyl)-4-fluoroaniline

PC1688F-100G PC1688F-25G PC1688F-5G
APOSPC1688F-100GEA 1440 DKK
APOSPC1688F-100G APOSPC1688F-25G APOSPC1688F-5G
2-Chlor-4'-fluoracetanilid 97%
2-Chlor-4'-fluoracetanilid
Formel: C₈H₇ClFNO
Smeltepunkt: 131…132 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00015730
CAS-nummer: 351-04-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR