O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: PFBHA.HCl

PC0810-1G PC0810-5G PC0810-25G
APOSPC0810-1GEA 491 DKK
APOSPC0810-1G APOSPC0810-5G APOSPC0810-25G
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride 97%
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride

Derivatising agent for Electron-Capture detection of steriods.

Formel: C₇H₄F₅NO
Smeltepunkt: 227 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012953
CAS-nummer: 57981-02-9
EINECS: 261-057-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR