4-(4-Morpholinyl)-1-butanol 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-Morpholinobutan-1-ol

OR963360-1G
APOSOR963360-1GEA 1630 DKK
APOSOR963360-1G
4-(4-Morpholinyl)-1-butanol 95%
4-(4-Morpholinyl)-1-butanol
Formel: C₈H₁₇NO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03211280
CAS-nummer: 5835-79-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR