4-(Hydroxymethyl)benzonitrile 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-Cyanobenzyl alcohol

OR962541-25G OR962541-100G
APOSOR962541-25GEA 763 DKK
APOSOR962541-25G APOSOR962541-100G
4-(Hydroxymethyl)benzonitrile 98%
4-(Hydroxymethyl)benzonitrile
Formel: C₈H₇NO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00870633
CAS-nummer: 874-89-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR