1-Cyclohexene-1-carboxylic acid 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Cyclohex-1-enecarboxylic acid

OR961865-25G
APOSOR961865-25GEA 2090 DKK
APOSOR961865-25G
1-Cyclohexene-1-carboxylic acid 95%
1-Cyclohexene-1-carboxylic acid
Formel: C₇H₁₀O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001545
CAS-nummer: 636-82-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR