3,3'-Bipiperidine 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR958208-1G OR958208-250MG
APOSOR958208-1GEA 3930 DKK
APOSOR958208-1G APOSOR958208-250MG
3,3'-Bipiperidine 95%
3,3'-Bipiperidine
Formel: C₁₀H₂₀N₂
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 46040-83-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR