1-(Methylamino)cyclohexanecarbonitrile 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR953808-1G OR953808-5G
APOSOR953808-1GEA 1640 DKK
APOSOR953808-1G APOSOR953808-5G
1-(Methylamino)cyclohexanecarbonitrile 95%
1-(Methylamino)cyclohexanecarbonitrile
Formel: C₈H₁₄N₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00972028
CAS-nummer: 6289-40-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR