2-Vinylphenyl acetate 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-Acetoxystyrene, Acetic Acid 2-Vinylphenyl Ester

OR942180-1G OR942180-250MG
APOSOR942180-1GEA 719 DKK
APOSOR942180-1G APOSOR942180-250MG
2-Vinylphenyl acetate 95%
2-Vinylphenyl acetate
Formel: C₁₀H₁₀O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02093430
CAS-nummer: 63600-35-1

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR