N-(2-Methoxyethyl)isopropylamine 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR937475-1G OR937475-5G
APOSOR937475-1GEA 295 DKK
APOSOR937475-1G APOSOR937475-5G
N-(2-Methoxyethyl)isopropylamine 95%
N-(2-Methoxyethyl)isopropylamine
Formel: C₆H₁₅NO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00144828
CAS-nummer: 104678-18-4

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR