2-tert-Butyl-6-methylaniline 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 6-tert-Butyl-o-toluidine

OR937298-1G
APOSOR937298-1GEA 3550 DKK
APOSOR937298-1G
2-tert-Butyl-6-methylaniline 95%
2-tert-Butyl-6-methylaniline
Formel: C₁₁H₁₇N
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00025208
CAS-nummer: 13117-94-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR