p-Toluidiniumchlorid 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: p-Toluidine hydrochloride, 4-Toluidine HCl, p-Toluidine HCl, 4-Aminotoluene hydrochloride, 4-Methylaniline HCl, 4-Methylaniline hydrochloride, 4-Aminotoluene HCl, 4-Toluidine hydrochloride

OR933321-100G OR933321-1KG OR933321-500G
APOSOR933321-100GEA 933 DKK
APOSOR933321-100G APOSOR933321-1KG APOSOR933321-500G
p-Toluidiniumchlorid 97%
p-Toluidiniumchlorid
Formel: C₇H₁₀ClN
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00035497
CAS-nummer: 540-23-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR