Methoxymethyl phenyl sulphide 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: alpha-Methoxythioanisole, Methyl Phenylthiomethyl Ether

OR932563-10G OR932563-1G OR932563-5G
APOSOR932563-10GEA 2180 DKK
APOSOR932563-10G APOSOR932563-1G APOSOR932563-5G
Methoxymethyl phenyl sulphide 95%
Methoxymethyl phenyl sulphide
Formel: C₈H₁₀OS
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008496
CAS-nummer: 13865-50-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR