2,2-Diethoxypropan 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Acetone diethyl Acetal

OR930301-25G OR930301-5G OR930301-100G
APOSOR930301-25GEA 2030 DKK
APOSOR930301-25G APOSOR930301-5G APOSOR930301-100G
2,2-Diethoxypropan 95%
2,2-Diethoxypropane
Formel: C₇H₁₆O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009224
CAS-nummer: 126-84-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR