4-(2-Acetoxyethoxy)toluene 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-(p-Tolyloxy)ethyl acetate, Ethylene Glycol Mono-p-tolyl Ether Acetate, Acetic Acid 2-p-Tolyloxyethyl Ester, 2-p-Tolyloxyethyl Acetate

OR919731-10G OR919731-1G OR919731-5G
APOSOR919731-10GEA 1010 DKK
APOSOR919731-10G APOSOR919731-1G APOSOR919731-5G
4-(2-Acetoxyethoxy)toluene 95%
4-(2-Acetoxyethoxy)toluene
Formel: C₁₁H₁₄O₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00059337
CAS-nummer: 6807-11-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR