2-Hydroxypropyl acrylate 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

OR919495-1G
APOSOR919495-1GEA 3010 DKK
APOSOR919495-1G
2-Hydroxypropyl acrylate 95%
2-Hydroxypropyl acrylate
Formel: C₆H₁₀O₃
Kogepunkt: 225 °C
Massefylde: 1,06 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 999-61-1
UN nummer: 2922
ADR: 8+6.1,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR