N,4-Dimethyl-2-nitroaniline 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR909293-1G OR909293-250MG
APOSOR909293-1GEA 4310 DKK
APOSOR909293-1G APOSOR909293-250MG
N,4-Dimethyl-2-nitroaniline 95%
N,4-Dimethyl-2-nitroaniline
Formel: C₈H₁₀N₂O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07437778
CAS-nummer: 4600-08-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR