1-Boc-3-(methylaminomethyl)azetidine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: tert-butyl 3-[(methylamino)methyl]azetidine-1-carboxylate, 3-[(Methylamino)methyl]azetidine, N1-BOC protected, 1-Boc-3-((Methylamino)methyl)azetidine

OR60182-250MG
APOSOR60182-250MGEA 1630 DKK
APOSOR60182-250MG
1-Boc-3-(methylaminomethyl)azetidine
1-Boc-3-(methylaminomethyl)azetidine
Formel: C₁₀H₂₀N₂O₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD08061965
CAS-nummer: 1049730-81-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR