Pyrimidine-5-carboxylic acid 97+%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR5339-1G OR5339-5G OR5339-10G
APOSOR5339-1GEA 164 DKK
APOSOR5339-1G APOSOR5339-5G APOSOR5339-10G
Pyrimidine-5-carboxylic acid 97+%
Pyrimidine-5-carboxylic acid
Formel: C5H4N2O2
Smeltepunkt: 259…261 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00856162
CAS-nummer: 4595-61-3
EINECS: 224-994-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR