3-Ethylamino-p-cresol 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Ethylamino-4-methylphenol

OR53165-25G
APOSOR53165-25GEA 712 DKK
APOSOR53165-25G
3-Ethylamino-p-cresol 95%
3-Ethylamino-p-cresol
Formel: C₉H₁₃NO
Smeltepunkt: 85…87 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002382
CAS-nummer: 120-37-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR