Terephthalaldehyde mono(diethyl acetal) 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-(Diethoxymethyl)benzaldehyde

OR4985-100G OR4985-25G
APOSOR4985-100GEA 1970 DKK
APOSOR4985-100G APOSOR4985-25G
Terephthalaldehyde mono(diethyl acetal) 97%
Terephthalaldehyde mono(diethyl acetal)
Formel: C₁₂H₁₆O₃
Kogepunkt: 89…93 °C/7mmHg
Massefylde: 1,047 g/cm³ (25 °C)
Flammepunkt: >113 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010217
CAS-nummer: 81172-89-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR