3-Methoxy-p-toluidin 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1-Amino-3-methoxy-4-methylbenzene, 4-Amino-2-methoxytoluene, 3-Methoxy-4-methylaniline, 4-Methyl-m-anisidine, 5-Amino-2-methylanisole

OR480640-25G OR480640-5G
APOSOR480640-25GEA 1560 DKK
APOSOR480640-25G APOSOR480640-5G
3-Methoxy-p-toluidin 95%
3-Methoxy-p-toluidin
Formel: C₈H₁₁NO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00025371
CAS-nummer: 16452-01-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR