2-(1H-Pyrrol-1-yl)-1,3,4-thiadiazole

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR46716-1G OR46716-250MG OR46716-500MG OR46716-2.5G OR46716-5G
APOSOR46716-1GEA 3300 DKK
APOSOR46716-1G APOSOR46716-250MG APOSOR46716-500MG APOSOR46716-2.5G APOSOR46716-5G
2-(1H-Pyrrol-1-yl)-1,3,4-thiadiazole
2-(1H-Pyrrol-1-yl)-1,3,4-thiadiazole
Formel: C₆H₅N₃S
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06653492
CAS-nummer: 881041-56-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR