(S)-1-Methyl-3-pyrrolidinol 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (S)-3-Hydroxy-1-methylpyrrolidine, (S)-(+)-1-Methyl-3-pyrrolidinol

OR4625-25G
APOSOR4625-25GEA 959 DKK
APOSOR4625-25G
(S)-1-Methyl-3-pyrrolidinol 97%
(S)-1-Methyl-3-pyrrolidinol

Chiral intermediate for pharmaceuticals and agrochemicals.

Formel: C₅H₁₁NO
Molekylvægt: 101.15 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03788748
CAS-nummer: 104641-59-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR