1-Bromo-2-iodoethane

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR460045-1G OR460045-5G
APOSOR460045-1GEA 1380 DKK
APOSOR460045-1G APOSOR460045-5G
1-Bromo-2-iodoethane
1-Bromo-2-iodoethane
Formel: C₂H₄BrI
Kogepunkt: 163 °C
Smeltepunkt: 28…32 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00041542
CAS-nummer: 590-16-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR