Acrolein diethyl acetal 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Acrolein Acetal, 1,1-Diethoxy-2-propene, 3,3-Diethoxy-1-propene

OR40659-100G OR40659-250G OR40659-25G
APOSOR40659-100GEA 600 DKK
APOSOR40659-100G APOSOR40659-250G APOSOR40659-25G
Acrolein diethyl acetal 98%
Acrolein diethyl acetal
Formel: C₇H₁₄O₂
Kogepunkt: 120…125 °C
Massefylde: 0,852 g/cm³ (25 °C)
Flammepunkt: 13 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009239
CAS-nummer: 3054-95-3
UN nummer: 2374
ADR: 3,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR