2-Hydroxy-4-pyridinboronsyrepinacolester

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR40294-1G OR40294-250MG OR40294-5G
APOSOR40294-1GEA 2050 DKK
APOSOR40294-1G APOSOR40294-250MG APOSOR40294-5G
2-Hydroxy-4-pyridinboronsyrepinacolester
2-Hydroxy-4-pyridinboronsyrepinacolester
Formel: C₁₁H₁₆BNO₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11223287
CAS-nummer: 1256358-90-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR