2-Brom-5-iodbenzoesyre

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR4005-1G OR4005-25G OR4005-5G
APOSOR4005-1GEA 164 DKK
APOSOR4005-1G APOSOR4005-25G APOSOR4005-5G
2-Brom-5-iodbenzoesyre
2-Brom-5-iodbenzoesyre
Formel: C₇H₄BrIO₂
Smeltepunkt: 171…175 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00079716
CAS-nummer: 25252-00-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR