2-Bromo-6-chlorophenol

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR399002-1G OR399002-5G
APOSOR399002-1GEA 164 DKK
APOSOR399002-1G APOSOR399002-5G
2-Bromo-6-chlorophenol
2-Bromo-6-chlorophenol
Formel: C₆H₄BrClO
Smeltepunkt: 55…56 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09907875
CAS-nummer: 2040-88-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR