2,6-Dimethyl-4-pyridinboronsyrepinacolester

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

OR360125-1G
APOSOR360125-1GEA 491 DKK
APOSOR360125-1G
2,6-Dimethyl-4-pyridinboronsyrepinacolester
2,6-Dimethyl-4-pyridinboronsyrepinacolester
Formel: C₁₃H₂₀BNO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11617853
CAS-nummer: 325142-95-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR