(2R,4R)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Boc-D-cis-Hyp-OMe, N-(tert-Butoxycarbonyl)-cis-4-hydroxy-D-proline methyll ester, 1-tert-Butyl 2-Methyl (2R,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylate, Boc-cis-D-Hyp-OMe, 1-tert-butyl 2-methyl (2R,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate, (2R,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylic acid 1-tert-Butyl 2-Methyl ester, (2R,4R)-1-tert-Butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, N-Boc-cis-4-hydroxy-D-proline methyll ester, N-Boc-cis-4-hydroxy-D-proline methyl ester

OR321377-100G
APOSOR321377-100GEA 1310 DKK
APOSOR321377-100G
(2R,4R)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate 97%
(2R,4R)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate
Formel: C₁₁H₁₉NO₅
Molekylvægt: 245.27 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD00797548
CAS-nummer: 114676-69-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR