Methyl-2-methyl-4-nitrobenzoate 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Methyl 2-methyl-4-nitrobenzoate

OR315322-100G
APOSOR315322-100GEA 806 DKK
APOSOR315322-100G
Methyl-2-methyl-4-nitrobenzoate 98%
Methyl-2-methyl-4-nitrobenzoate
Formel: C₉H₉NO₄
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03618407
CAS-nummer: 62621-09-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR