1-[3-(3-Methyl-1,2,4-oxadiazol-5-yl)pyridin-2-yl]-1H-imidazole-4-carboxylic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR310707-1G
APOSOR310707-1GEA 5860 DKK
APOSOR310707-1G
1-[3-(3-Methyl-1,2,4-oxadiazol-5-yl)pyridin-2-yl]-1H-imidazole-4-carboxylic acid
1-[3-(3-Methyl-1,2,4-oxadiazol-5-yl)pyridin-2-yl]-1H-imidazole-4-carboxylic acid
Formel: C₁₂H₉N₅O₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16629529
CAS-nummer: 1239851-29-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR