cis-tert-Butyl-3-hydroxycyclobutylcarbamate 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: tert-Butyl (cis-3-hydroxycyclobutyl)carbamate

OR308102-1G OR308102-5G OR308102-25G
APOSOR308102-1GEA 632 DKK
APOSOR308102-1G APOSOR308102-5G APOSOR308102-25G
cis-tert-Butyl-3-hydroxycyclobutylcarbamate 97%
cis-tert-Butyl-3-hydroxycyclobutylcarbamate
Formel: C₉H₁₇NO₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09038208
CAS-nummer: 389890-43-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR