3-tert-Butoxycarbonylamino-4-ethyl-hexanoic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR307570-1G
APOSOR307570-1GEA 3510 DKK
APOSOR307570-1G
3-tert-Butoxycarbonylamino-4-ethyl-hexanoic acid
3-tert-Butoxycarbonylamino-4-ethyl-hexanoic acid
Formel: C₁₃H₂₅NO₄
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD05863634
CAS-nummer: 776330-51-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR