4-Brom-2-nitroanilin

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR3065-100G OR3065-250G OR3065-500G
APOSOR3065-100GEA 349 DKK
APOSOR3065-100G APOSOR3065-250G APOSOR3065-500G
4-Brom-2-nitroanilin
4-Brom-2-nitroanilin
Formel: C₆H₅BrN₂O₂
Smeltepunkt: 110…114 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00041312
CAS-nummer: 875-51-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR