4-Nitro-1-isoindolinone 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Nitroisoindolin-1-one

OR300701-1G OR300701-250MG OR300701-5G
APOSOR300701-1GEA 818 DKK
APOSOR300701-1G APOSOR300701-250MG APOSOR300701-5G
4-Nitro-1-isoindolinone 98%
4-Nitro-1-isoindolinone
Formel: C₈H₆N₂O₃
Smeltepunkt: 225…230 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09701290
CAS-nummer: 366452-97-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR