3-(2,4-Dichlorphenyl)propionsyre

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3-(2,4-Dichlorophenyl)propanoic acid

OR29293-25G
APOSOR29293-25GEA 959 DKK
APOSOR29293-25G
3-(2,4-Dichlorphenyl)propionsyre
3-(2,4-Dichlorphenyl)propionsyre
Formel: C₉H₈Cl₂O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00275605
CAS-nummer: 55144-92-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR