4-tert-Butyl-o-phenylendiamin

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-(tert-Butyl)benzene-1,2-diamine, 4-tert-Butylbenzene-1,2-diamine, 4-tert-Butyl-1,2-phenylenediamine, 1,2-Diamino-4-(dimethylethyl)benzene, 1,2-Diamino-4-tert-butylbenzene

OR27181-1G OR27181-250MG OR27181-5G
APOSOR27181-1GEA 218 DKK
APOSOR27181-1G APOSOR27181-250MG APOSOR27181-5G
4-tert-Butyl-o-phenylendiamin
4-tert-Butyl-o-phenylendiamin
Formel: C₁₀H₁₆N₂
Smeltepunkt: 96…100 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00052695
CAS-nummer: 68176-57-8
UN nummer: 2811
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR