Cyclopropyl phenyl sulphide

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Cyclopropyl(phenyl)sulphane

OR18541-1G
APOSOR18541-1GEA 164 DKK
APOSOR18541-1G
Cyclopropyl phenyl sulphide
Cyclopropyl phenyl sulphide
Formel: C₉H₁₀S
Kogepunkt: 62…63 °C/1mmHg
Massefylde: 1,045 g/cm³ (25 °C)
Flammepunkt: 92 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009684
CAS-nummer: 14633-54-6
UN nummer: 2810
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR