Choline-p-toluenesulphonate 85%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (2-Hydroxyethyl)trimethylammonium toluene-4-sulphonate, 2-Hydroxy-N,N,N-trimethylethanaminium 4-methylbenzenesulphonate

OR18384-5G OR18384-25G
APOSOR18384-5GEA 273 DKK
APOSOR18384-5G APOSOR18384-25G
Choline-p-toluenesulphonate 85%
Choline-p-toluenesulphonate
Formel: C₅H₁₄NO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00079051
CAS-nummer: 55357-38-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR